SC-514

Synonyms: 4-amino-[2,3'-bithiophene]-5-carboxamideIKK-2 Inhibitor, SC-514

2442-25 2442-5
10189-926EA 344.42 USD
10189-926 10189-924
SC-514
SC-514
Formula: C₉H₈N₂OS₂
MW: 224.3 g/mol
MDL Number: MFCD00206459
CAS Number: 354812-17-2

Order Now

Specification Test Results

CAS 354812-17-2
Purity ≥98% by TLC
Form Yellow solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR