Mdivi -1

Synonyms: 3-(2,4-Dichloro-5-methoxyphenyl)-2-mercaptoquinazolin-4(3H)-one3-(2,4-Dichloro-5-methoxyphenyl)-2-thioxo-2,3-dihydro-4(1H)-quinazolinone

2441-25 2441-5
10189-922EA 344.42 USD
10189-922 10189-920
Mdivi -1
Mdivi -1
Formula: C₁₅H₁₀Cl₂N₂O₂S
MW: 353.2 g/mol
MDL Number: MFCD00974506
CAS Number: 338967-87-6

Order Now

Specification Test Results

CAS 338967-87-6
Purity ≥98% by TLC
Form White solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR