Daminozide (succinic acid mono(2,2-dimethylhydrazide))

Synonyms: DaminozideSuccinic acid 2,2-Dimethylhydrazide

2438-100 2438-500
10192-040EA 75.89 USD
10192-040 10191-370
Daminozide (succinic acid mono(2,2-dimethylhydrazide))
Daminozide (Succinic acid mono(2,2-dimethylhydrazide))

Order Now

Specification Test Results

CAS 1596-84-5
Purity ≥95%
Form White solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR