CEP-18770

Synonyms: ((R)-1-((2S,3R)-3-Hydroxy-2-(6-phenylpicolinamido)butanamido)-3-methylbutyl)boronic acid

2433-25 2433-5
10189-910EA 227.66 USD
10189-910 10189-912
CEP-18770
CEP-18770
Formula: C₂₁H₂₈BN₃O₅
MW: 413,28 g/mol
MDL Number: MFCD18251439
CAS Number: 847499-27-8

Order Now

Specification Test Results

CAS 847499-27-8
Purity ≥98% by HPLC
Form Off-white solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR