TPCA-1

Synonyms: 5-(4-Fluorophenyl)-2-ureidothiophene-3-carboxamideIKK2 Inhibitor IV2-[(aminocarbonyl)amino]-5-(4-fluorophenyl)-3-thiophenecarboxamide

2431-5 2431-25
10189-962EA 134.06 USD
10189-962 10191-846
TPCA-1
TPCA-1
Formula: C₁₂H₁₀FN₃O₂S
MW: 279.3 g/mol
MDL Number: MFCD09037541
CAS Number: 507475-17-4

Order Now

Specification Test Results

CAS 507475-17-4
Purity ≥95% by HPLC
Form Off-white solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR