Semagacestat

Synonyms: (S)-2-Hydroxy-3-methyl-N-((S)-1-(((S)-3-methyl-2-oxo-2,3,4,5-tetrahydro-1H-benzo[d]azepin-1-yl)amino)-1-oxopropan-2-yl)butanamide

2430-5 2430-1
10189-960EA 344.42 USD
10189-960 10189-958
Semagacestat
Semagacestat
Formula: C₁₉H₂₇N₃O₄
MW: 361,44 g/mol
MDL Number: MFCD25542369
CAS Number: 425386-60-3

Order Now

Specification Test Results

CAS 425386-60-3
Purity ≥98% by HPLC
Form White solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR