Anacetrapib
Synonyms:
(4S,5R)-5-(3,5-Bis(trifluoromethyl)phenyl)-3-((4'-fluoro-5'-isopropyl-2'-methoxy-4-(trifluoromethyl)-[1,1'-biphenyl]-2-yl)methyl)-4-methyloxazolidin-2-one
2418-5
2418-1
10189-892EA
332.22
USD
10189-892
10189-890
Anacetrapib
Anacetrapib
Formula:
C₃₀H₂₅F₁₀NO₃ MW: 637,51 g/mol |
MDL Number:
MFCD16294903 CAS Number: 875446-37-0 |
Specification Test Results
CAS | 875446-37-0 |
Purity | ≥98% |
Form | Crystalline solid |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...