Phloridzin

Synonyms: Phlorizin hydrate

2416-100 2416-500
10191-844EA 227.32 USD
10191-844 10191-842
Phloridzin
Phloridzin
Formula: C₂₁H₂₄O₁₀
MW: 436.41 g/mol
MDL Number: MFCD00006591
CAS Number: 60-81-1

Order Now

Specification Test Results

CAS 60-81-1
Purity ≥98%
Form White to yellow solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR