LFM-A13
2414-5
2414-25
10189-882EA
437.81
USD
10189-882
10189-880
LFM-A13
LFM-A13
CAS Number:
244240-24-2 |
Specification Test Results
CAS | 244240-24-2 |
Purity | ≥98% by TLC |
Form | White solid |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...