JNJ-1661010
2413-5
2413-25
10191-364EA
437.81
USD
10191-364
10191-362
JNJ-1661010
JNJ-1661010
CAS Number:
681136-29-8 |
Specification Test Results
CAS | 681136-29-8 |
Purity | ≥98% by TLC |
Form | Off-white solid |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...