GW5074

Synonyms: 5-Iodo-3-[(3,5-dibromo-4-hydroxyphenyl)methylene]-2-indolinone3-(3,5-dibromo-4-hydroxybenzylidene)-5-iodoindolin-2-one

2412-5 2412-25
10191-360EA 531.05 USD
10191-360 10189-878
GW5074
GW5074
Formula: C₁₅H₈Br₂INO₂
MW: 520.94 g/mol
MDL Number: MFCD09971042
CAS Number: 220904-83-6

Order Now

Specification Test Results

CAS 220904-83-6
Purity ≥98% by TLC
Form Dark orange solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR