(±)-Blebbistatin

2405-5 2405-25
10189-852EA 174.19 USD
10189-852 10189-854
(±)-Blebbistatin
(±)-Blebbistatin
Formula: C₁₈H₁₆N₂O₂
MW: 292.33 g/mol
CAS Number: 674289-55-5

Order Now

Specification Test Results

CAS 674289-55-5
Purity ≥98% by HPLC
Form Yellow solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR