FCCP

Synonyms: [4-(Trifluoromethoxy)phenylhydrazono]malononitrile[4-(Trifluoromethoxy)phenyl]carbonohydrazonoyl dicyanideFCCPCarbonyl cyanide 4-(Trifluoromethoxy)phenylhydrazone

2398-25 2398-5
10189-834EA 355.97 USD
10189-834 10189-832
FCCP
FCCP
Formula: C₁₀H₅F₃N₄O
MW: 254.17 g/mol
MDL Number: MFCD00009699
CAS Number: 370-86-5

Order Now

Specification Test Results

CAS 370-86-5
Purity ≥98% by TLC
Form Pale yellow solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR