CL-82198 (MMP-13 inhibitor)

Synonyms: N-[4-(4-morpholinyl)butyl]-2-benzofurancarboxamide

2392-25 2392-5
10189-810EA 437.81 USD
10189-810 10189-808
CL-82198 (MMP-13 inhibitor)
CL-82198 (MMP-13 inhibitor)
Formula: C₁₇H₂₂N₂O₃
MW: 302.37 g/mol
MDL Number: MFCD12828877
CAS Number: 307002-71-7

Order Now

Specification Test Results

CAS 307002-71-7
Purity ≥98% by TLC
Form Off-white solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR