Doxorubicin hydrochloride ≥95% (by TLC)

Synonyms: (8S,10S)-10-(((2R,4S,5S,6S)-4-Amino-5-hydroxy-6-methyltetrahydro-2H-pyran-2-yl)oxy)-6,8,11-trihydroxy-8-(2-hydroxyacetyl)-1-methoxy-7,8,9,10-tetrahydrotetracene-5,12-dione hydrochlorideHydroxydaunorubicin Hydrochloride

1527-5
10187-480EA 125.18 USD
10187-480
Doxorubicin hydrochloride ≥95% (by TLC)
Doxorubicin hydrochloride

Doxorubicin.hydrochloride is an anti-tumor antibiotic and an immunosuppressive agent that inhibits reverse transcriptase and RNA polymerase.


  • Orange red powder


This is a highly effective myotoxin that inhibits topoisomerase II and binds to Nucleic acids, presumably by specific intercalation into the DNA double helix, thereby inhibiting necleic acid synthesis. Induces apoptosis.


Soluble in H2O


Caution: For research use only, not for use in humans.

Formula: C₂₇H₂₉NO₁₁·HCl
MW: 579.98 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00077757
CAS Number: 25316-40-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR