Oprozomib

Synonyms: N-((S)-3-Methoxy-1-(((S)-3-methoxy-1-(((S)-1-((R)-2-methyloxiran-2-yl)-1-oxo-3-phenylpropan-2-yl)amino)-1-oxopropan-2-yl)amino)-1-oxopropan-2-yl)-2-methylthiazole-5-carboxamide

2386-25 2386-5
10189-796EA 135.88 USD
10189-796 10189-798
Oprozomib
Oprozomib
Formula: C₂₅H₃₂N₄O₇S
MW: 532,61 g/mol
MDL Number: MFCD25976563
CAS Number: 935888-69-0

Order Now

Specification Test Results

CAS 935888-69-0
Purity ≥98% by HPLC
Form White to off-white solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR