Daunorubicin hydrochloride ≥95%

Supplier: BioVision

Synonyms: Hydroxydaunorubicin Hydrochloride

1524-500 AB141394-50MG AB141394-10MG
10187-478EA 1774.6 USD
10187-478 10187-476 10187-474
Daunorubicin hydrochloride ≥95%
Daunorubicin hydrochloride

Potent anti-cancer agent whose potential target site may be mitochondrial cytochrome-c-oxidase


  • Orange red powder


Inhibits RNA and DNA synthesis. Also inhibits eukaryotic topoisomerase I & II. Induces DNA single strand breaks and apoptosis in HeLaS3 tumor cells.


Soluble in H2O or MeOH


Caution: For research use only, not for use in humans.

Formula: C₂₇H₂₉NO₁₁·HCl
MW: 563.98 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD04974507
CAS Number: 23541-50-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR