Diphenyleneiodonium chloride
Synonyms:
Diphenyleneiodonium Chloride
2358-5
10189-708EA
87.57
USD
10189-708
Diphenyleneiodonium chloride
Diphenyleneiodonium chloride
Specification Test Results
CAS | 4673-26-1 |
Purity | ≥98% |
Form | White solid |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...