Torin 2, EZSolution™

Synonyms: 9-(6-Aminopyridin-3-yl)-1-(3-(trifluoromethyl)phenyl)benzo[h][1,6]naphthyridin-2(1H)-one

2354-5
10189-694EA 145.89 USD
10189-694
Torin 2, EZSolution™
Torin 2
Formula: C₂₄H₁₅F₃N₄O
MW: 432,4 g/mol
MDL Number: MFCD18782652
CAS Number: 1223001-51-1

Order Now

Specification Test Results

CAS 1223001-51-1
Purity ≥98% by HPLC
Form Liquid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR