Torin 1, EZSolution™

Synonyms: 1-(4-(4-Propionylpiperazin-1-yl)-3-(trifluoromethyl)phenyl)-9-(quinolin-3-yl)benzo[h][1,6]naphthyridin-2(1H)-one

2353-5
10191-820EA 175.56 USD
10191-820
Torin 1, EZSolution™
Torin 1
Formula: C₃₅H₂₈F₃N₅O₂
MW: 607,62 g/mol
MDL Number: MFCD18782653
CAS Number: 1222998-36-8

Order Now

Specification Test Results

CAS 1222998-36-8
Purity ≥98% by HPLC
Form Liquid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR