AS-604850

Synonyms: 5-((2,2-Difluorobenzo[d][1,3]dioxol-5-yl)methylene)thiazolidine-2,4-dione

2339-5 2339-1
10189-636EA 70.04 USD
10189-636 10189-638
AS-604850
AS-604850
Formula: C₁₁H₅F₂NO₄S
MW: 285.2 g/mol
MDL Number: MFCD19705528
CAS Number: 648449-76-7

Order Now

Specification Test Results

CAS 648449-76-7
Purity ≥98% by HPLC
Form Off-white solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR