10-NCP

2338-1 2338-5
10189-632EA 99.08 USD
10189-632 10189-634
10-NCP
10-NCP
Formula: C₂₀H₂₅ClN₂O·HCl
MW: 381.34 g/mol
MDL Number: MFCD09038570
CAS Number: 925681-41-0

Order Now

Specification Test Results

CAS 925681-41-0
Purity ≥98% by HPLC
Form White solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR