Laquinimod
2332-5
2332-25
10189-620EA
169.28
USD
10189-620
10189-622
Laquinimod
Laquinimod
Formula:
C₁₉H₁₆ClN₂NaO₃ |
CAS Number:
248282-07-7 |
Specification Test Results
CAS | 248282-07-7 |
Purity | ≥98% by HPLC |
Form | Off-white solid |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...