N-Hydroxysuccinimide
Synonyms:
1-Hydroxypyrrolidine-2,5-dione
, NHS
, HOSu
2331-25G
2331-5G
10189-618EA
227.66
USD
10189-618
10189-616
N-Hydroxysuccinimide
N-Hydroxysuccinimide
Formula:
C₄H₅NO₃ MW: 115.09 g/mol |
MDL Number:
MFCD00005516 CAS Number: 6066-82-6 |
Specification Test Results
CAS | 6066-82-6 |
Purity | ≥97% by GC |
Form | White solid |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...