BS3 (Bis(sulfosuccinimidyl) suberate sodium salt) ≥90% (by NMR) crosslinker
Supplier: BioVision
Synonyms:
BS³
, Sulfo DSS
, bis(sulphosuccinimidyl)suberate
, Suberic acid bis(3-sulfo-N-hydroxysuccinimide ester) sodium salt
, bis(sulfosuccinimidyl)suberate
, Suberic acid bis(3-sulpho-N-hydroxysuccinimide ester) sodium salt
A water-soluble, homobifunctional protein crosslinker.
- Membrane-impermeable
- Protect from moisture
- Purity: ≥90% by NMR
- Solubility: water (>6 mg/ml)
Amine-reactive and have a non-cleavable spacer with arm length of 11.4 Å. Crosslinking of proteins through reaction with primary amines. Useful for cross-linking cell surface proteins due to the membrane-impermeability of this reagent.
Caution: For research use only, not for use in humans.
Formula:
C₁₆H₁₈N₂Na₂O₁₄S₂ MW: 572.43 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00054982 CAS Number: 82436-77-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...