PJ-34 hydrochloride

Synonyms: 2-(Dimethylamino)-N-(6-oxo-5,6-dihydrophenanthridin-2-yl)acetamide hydrochloride

2319-5 2319-1
10189-558EA 70.04 USD
10189-558 10189-560
PJ-34 hydrochloride
PJ-34 hydrochloride
Formula: C₁₇H₁₇N₃O₂·HCl
MW: 295.2·36.5 g/mol
MDL Number: MFCD22581510
CAS Number: 344458-15-7

Order Now

Specification Test Results

CAS 344458-15-7
Purity ≥98% by HPLC
Form Pale yellow solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR