Ionomycin free acid ≥98%, EZSolution™

2309-5
10189-528EA 426.14 USD
10189-528
Ionomycin free acid ≥98%, EZSolution™
Ionomycin

This antibiotic acts as a potent and selective calcium ionophore.


  • Liquid


Ionomycin binds Ca2+ in the 7.0 to 9.5 pH range with the resulting complex exhibiting intense UV absorption. It induces apoptosis in immature B cell lines and in cultured embryonic rat cortical neurons. More mature B cell lines do not undergo apoptosis, but are arrested in the G1 phase of the cell cycle.


Caution: For research use only, not for use in humans.

Formula: C₄₁H₇₂O₉
Storage Temperature: Freezer
MDL Number: MFCD06798385
CAS Number: 56092-81-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR