Disulfiram

Synonyms: Bis(diethylthiocarbamoyl) disulphideTetraethylthiuram disulfideBis(diethylthiocarbamoyl) disulfideTetraethylthiuram disulphide1,1',1'',1'''-{Disulphanediylbis[(thioxomethylene)-nitrilo]}tetraethane1,1',1'',1'''-{Disulfanediylbis[(thioxomethylene)-nitrilo]}tetraethane

2308-250 2308-50
10189-526EA 192.64 USD
10189-526 10189-524
Disulfiram
Disulphiram
Formula: C₁₀H₂₀N₂S₄
MW: 296.55 g/mol
MDL Number: MFCD00009048
CAS Number: 97-77-8

Order Now

Specification Test Results

CAS 97-77-8
Purity ≥97% by titration
Form Pale yellow solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR