L(-)-Glutathione (oxidised form) ≥95%

Supplier: BioVision

Synonyms: L(-)-Glutathione, oxidizedGSSGBi(glutathion-S-yl)Glutathione oxidizedGlutathiolOxidized Glutathione (GSSG)

1241-1
10187-458EA 115.95 USD InStock
10187-458
L(-)-Glutathione (oxidised form) ≥95%
L(-)-Glutathione (oxidized form)
Formula: C₂₀H₃₂N₆O₁₂S₂
MW: 612,64 g/mol
Melting Pt: 185 °C
Storage Temperature: 2 - 8°C
MDL Number: MFCD00063106
CAS Number: 27025-41-8
EINECS: 248-170-7
UN: 0000

Order Now

Specification Test Results

CAS 27025-41-8
Purity 95%
Form White solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR