Caffeic acid

Synonyms: 3,4-Dihydroxycinnamic acid3-(3,4-Dihydroxyphenyl)acrylic acid

2303-50 2303-250
10189-504EA 52.54 USD
10189-504 10189-506
Caffeic acid
Caffeic acid
Formula: C₉H₈O₄
MW: 180.16 g/mol
MDL Number: MFCD00004392
CAS Number: 331-39-5

Order Now

Specification Test Results

CAS 331-39-5
Purity ≥98% by HPLC
Form Yellowish brown solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR