PR-619

Synonyms: 3,5-Dithiocyanatopyridine-2,6-diamine

2302-25 2302-5
10189-500EA 111.89 USD
10189-500 10189-502
PR-619
PR-619
Formula: C₇H₅N₅S₂
MW: 223.2781 g/mol
MDL Number: MFCD00830384
CAS Number: 2645-32-1

Order Now

Specification Test Results

CAS 2645-32-1
Purity ≥98% by HPLC
Form Yellow solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR