P 22077

Synonyms: 1-(5-((2,4-Difluorophenyl)thio)-4-nitrothiophen-2-yl)ethanone

2301-5 2301-25
10189-498EA 344.42 USD
10189-498 10189-496
P 22077
P 22077
Formula: C₁₂H₇F₂NO₃S₂
MW: 315,32 g/mol
MDL Number: MFCD22580421
CAS Number: 1247819-59-5

Order Now

Specification Test Results

CAS 1247819-59-5
Purity ≥98% by HPLC
Form Yellow solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR