OSU-03012

Synonyms: 2-Amino-N-(4-(5-(phenanthren-2-yl)-3-(trifluoromethyl)-1H-pyrazol-1-yl)phenyl)acetamide

2295-25 2295-5
10189-474EA 391.11 USD
10189-474 10189-472
OSU-03012
OSU-03012
Formula: C₂₆H₁₉F₃N₄O
MW: 460,45 g/mol
MDL Number: MFCD12912272
CAS Number: 742112-33-0

Order Now

Specification Test Results

CAS 742112-33-0
Purity ≥98% by HPLC
Form White to off-white solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR