BIIB 021

Synonyms: 6-Chloro-9-((4-methoxy-3,5-dimethylpyridin-2-yl)methyl)-9H-purin-2-amine

2291-5 2291-25
10189-464EA 379.44 USD
10189-464 10189-462
BIIB 021
BIIB 021
Formula: C₁₄H₁₅ClN₆O
MW: 318,76 g/mol
MDL Number: MFCD15528939
CAS Number: 848695-25-0

Order Now

Specification Test Results

CAS 848695-25-0
Purity ≥99% by HPLC
Form White to off-white solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR