PF-04691502

Synonyms: 2-Amino-8-(rel-(1r,4r)-4-(2-hydroxyethoxy)cyclohexyl)-6-(6-methoxypyridin-3-yl)-4-methylpyrido[2,3-d]pyrimidin-7(8H)-one

2289-25 2289-5
10189-456EA 811.42 USD
10189-456 10189-454
PF-04691502
PF-04691502
Formula: C₂₂H₂₇N₅O₄
MW: 425,48 g/mol
MDL Number: MFCD18782794
CAS Number: 1013101-36-4

Order Now

Specification Test Results

CAS 1013101-36-4
Purity ≥98% by HPLC
Form Crystalline solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR