GSK1349572

Synonyms: Sodium (4R,12aS)-9-((2,4-difluorobenzyl)carbamoyl)-4-methyl-6,8-dioxo-3,4,6,8,12,12a-hexahydro-2H-pyrido[1',2':4,5]pyrazino[2,1-b][1,3]oxazin-7-olate

2288-25 2288-5
10189-452EA 906.43 USD
10189-452 10189-450
GSK1349572
GSK1349572
Formula: C₂₀H₁₈F₂N₃NaO₅
MW: 441,36 g/mol
MDL Number: MFCD28405599
CAS Number: 1051375-19-9

Order Now

Specification Test Results

CAS 1051375-16-6
Purity ≥98% by HPLC
Form Crystalline solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR