GSK461364

Synonyms: (R)-5-(6-((4-Methylpiperazin-1-yl)methyl)-1H-benzo[d]imidazol-1-yl)-3-(1-(2-(trifluoromethyl)phenyl)ethoxy)thiophene-2-carboxamide

2287-25 2287-5
10189-448EA 904.81 USD
10189-448 10189-446
GSK461364
GSK461364
Formula: C₂₇H₂₈F₃N₅O₂S
MW: 543,6 g/mol
MDL Number: MFCD12755419
CAS Number: 929095-18-1

Order Now

Specification Test Results

CAS 929095-18-1
Purity ≥98% by HPLC
Form Crystalline solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR