GSK1904529A

Synonyms: N-(2,6-Difluorophenyl)-5-(3-(2-((5-ethyl-2-methoxy-4-(4-(4-(methylsulphonyl)piperazin-1-yl)piperidin-1-yl)phenyl)amino)pyrimidin-4-yl)imidazo[1,2-a]pyridin-2-yl)-2-methoxybenzamideN-(2,6-Difluorophenyl)-5-(3-(2-((5-ethyl-2-methoxy-4-(4-(4-(methylsulfonyl)piperazin-1-yl)piperidin-1-yl)phenyl)amino)pyrimidin-4-yl)imidazo[1,2-a]pyridin-2-yl)-2-methoxybenzamide

2286-5 2286-25
10189-444EA 904.81 USD
10189-444 10189-442
GSK1904529A
GSK1904529A
Formula: C₄₄H₄₇F₂N₉O₅S
MW: 851,96 g/mol
MDL Number: MFCD17010271
CAS Number: 1089283-49-7

Order Now

Specification Test Results

CAS 1089283-49-7
Purity ≥98% by HPLC
Form Crystalline solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR