SB 939

Synonyms: (E)-3-(2-Butyl-1-(2-(diethylamino)ethyl)-1H-benzo[d]imidazol-5-yl)-N-hydroxyacrylamide

2285-5 2285-1
10189-438EA 75.89 USD
10189-438 10189-440
SB 939
SB 939
Formula: C₂₀H₃₀N₄O₂
MW: 358,48 g/mol
MDL Number: MFCD17215206
CAS Number: 929016-96-6

Order Now

Specification Test Results

CAS 929016-96-6
Purity ≥98%
Form Crystalline solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR