P5091

Synonyms: 1-(5-((2,3-Dichlorophenyl)thio)-4-nitrothiophen-2-yl)ethan-1-one

2277-25 2277-5
10189-410EA 344.42 USD
10189-410 10189-408
P5091
P5091
Formula: C₁₂H₇Cl₂NO₃S₂
MW: 348,22 g/mol
MDL Number: MFCD00123202
CAS Number: 882257-11-6

Order Now

Specification Test Results

CAS 882257-11-6
Purity ≥98% by TLC
Form White solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR