ABTS (Diammonium 2,2'-azinobis[3-ethyl-2,3-dihydrobenzothiazole-6-sulfonate])

Supplier: BioVision

Synonyms: Ammonium 2,2'-(hydrazine-1,2-diylidene)bis(3-ethyl-2,3-dihydrobenzo[d]thiazole-6-sulfonate)2,2′-Azino-bis(3-ethylbenzthiazoline-6-sulphonic acid) diammonium salt2,2′-Azino-bis(3-ethylbenzthiazoline-6-sulfonic acid) diammonium salt

AB142041-100ML
10187-452EA 230.04 USD
10187-452
ABTS (Diammonium 2,2'-azinobis[3-ethyl-2,3-dihydrobenzothiazole-6-sulfonate])
ABTS (Diammonium 2,2'-azinobis[3-ethyl-2,3-dihydrobenzothiazole-6-sulphonate])
Formula: C₁₈H₂₄N₆O₆S₄
MW: 548.68 g/mol
MDL Number: MFCD00010404
CAS Number: 30931-67-0

Order Now

Specification Test Results

CAS 30931-67-0
Form Liquid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR