KT5720

2276-50 2276-100
10189-404EA 145.72 USD
10189-404 10189-406
KT5720
KT5720
CAS Number: 108068-98-0

Order Now

Specification Test Results

CAS 108068-98-0
Purity ≥98% by HPLC
Form Colorless solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR