Cyclic pifithrin-alpha-hydrobromide

Synonyms: 2-(p-Tolyl)-5,6,7,8-tetrahydroimidazo[2,1-b]benzothiazole Hydrobromide2-(p-Tolyl)-5,6,7,8-tetrahydrobenzo[d]imidazo[2,1-b]thiazole hydrobromide

2270-5
10189-384EA 192.64 USD
10189-384
Cyclic pifithrin-alpha-hydrobromide
Cyclic pifithrin-alpha-hydrobromide
Formula: C₁₆H₁₆N₂S
MW: 349.29 g/mol
MDL Number: MFCD02683960
CAS Number: 511296-88-1

Order Now

Specification Test Results

CAS 511296-88-1
Purity ≥98% by HPLC
Form White solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR