KY02111

Synonyms: N-(6-Chloro-2-benzothiazolyl)-3,4-dimethoxybenzenepropanamideN-(6-Chlorobenzo[d]thiazol-2-yl)-3-(3,4-dimethoxyphenyl)propanamide

2264-25 2264-5
10189-360EA 75.89 USD
10189-360 10189-362
KY02111
KY02111
Formula: C₁₈H₁₇ClN₂O₃S
MW: 376,86 g/mol
MDL Number: MFCD08483062
CAS Number: 1118807-13-8

Order Now

Specification Test Results

CAS 1118807-13-8
Purity ≥98% by HPLC
Form Pale yellow solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR