WYE-23
2257-1000
2257-500
10189-334EA
128.42
USD
10189-334
10189-332
WYE-23
WYE-23
MW:
520.6 g/mol |
CAS Number:
1062169-46-3 |
Specification Test Results
CAS | 1062169-46-3 |
Purity | ≥98% by HPLC |
Form | White solid |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...