WYE-125132
2256-500
2256-1000
10189-328EA
81.72
USD
10189-328
10189-330
WYE-125132
WYE-125132
MW:
519.6 g/mol |
CAS Number:
1144068-46-1 |
Specification Test Results
CAS | 1144068-46-1 |
Purity | ≥98% by HPLC |
Form | White solid |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...