Binimetinib

Synonyms: MEK-162

2254-5 2254-25
10191-808EA 286.04 USD
10191-808 10191-806
Binimetinib
Binimetinib
Formula: C₁₇H₁₅BrF₂N₄O₃
MW: 441,23 g/mol
MDL Number: MFCD22124525
CAS Number: 606143-89-9

Order Now

Specification Test Results

CAS 606143-89-9
Purity ≥98% by HPLC
Form White to off-white solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR