PCI 34051

Supplier: BioVision

Synonyms: N-Hydroxy-1-(4-methoxybenzyl)-1H-indole-6-carboxamide

AB300227-25MG AB300227-5MG
10189-322EA 574.58 USD
10189-322 10189-320
PCI 34051
PCI 34051
Formula: C₁₇H₁₆N₂O₃
MW: 296,32 g/mol
MDL Number: MFCD11114393
CAS Number: 950762-95-5

Order Now

Specification Test Results

CAS 950762-95-5
Purity ≥95% by HPLC
Form White to Off-white Solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR