Clofarabine

Synonyms: 2-Chloro-2'-arabino-fluoro-2'-deoxyadenosine(2R,3R,4S,5R)-5-(6-Amino-2-chloro-9H-purin-9-yl)-4-fluoro-2-(hydroxymethyl)tetrahydrofuran-3-ol2-Chloro-9-(2-deoxy-2-fluoro-beta-D-arabinofuranosyl)adenine

2236-5 2236-25
10189-268EA 75.89 USD
10189-268 10189-270
Clofarabine
Clofarabine
Formula: C₁₀H₁₁ClFN₅O₃
MW: 303.68 g/mol
MDL Number: MFCD00871077
CAS Number: 123318-82-1

Order Now

Specification Test Results

CAS 123318-82-1
Purity ≥99% by HPLC
Form Off-white solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR