Necrox-5™ methanesulfonate

Synonyms: [5-(1,1-Dioxo-thiomorpholin-4-ylmethyl)-2-phenyl-1H-indol-7-yl]-(tetrahydro-pyran-4-ylmethyl)-amine

2229-1 2229-5
10189-250EA 151.54 USD
10189-250 10189-252
Necrox-5™ methanesulfonate
Necrox-5™ methanesulfonate
Formula: C₂₅H₃₁N₃O₃S·2CH₄O₃S
MW: 645.8 g/mol
CAS Number: 1383718-29-3

Order Now

Specification Test Results

Purity ≥95% by HPLC
Form Light pink solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR